
2-Bromo-3-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3430-17-9
:
Formula:C₆H₆BrN
MDL Number:MFCD00239380
:172.02 g/mol
Boiling Pt:218…219 °C
Density:1.544 g/cm³ (25 °C)
Flash Pt:113 °C
:GHS07
Storage Temperature:Ambient