
4-Amino-2-methylpyrimidine-5-carbonitrile
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:698-29-3
:
Formula:C₆H₆N₄
MDL Number:MFCD00084875
:134.14 g/mol
Melting Pt:249…252 °C
Storage Temperature:Ambient
EINECS:211-814-2
UN:3439
ADR:6.1,III
:GHS07