Order Entry
Finland
ContactUsLinkComponent
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Flammable
Toxic
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Catalog # APOSOR40785-10G
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Toxic
1-Acetoxy-1,3-butadiene (mixture of cis and trans isomeres) 90%
Catalog # APOSOR40785-10G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
Storage Temperature:Refrigerator
UN:1992
ADR:3+6.1,III
 :GHS02
 :GHS06
CAS Number:1515-76-0
Formula:C₆H₈O₂
MDL Number:MFCD00008714