Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Hydroxy-m-toluic acid
2-Hydroxy-m-toluic acid
2-Hydroxy-m-toluic acid
Irritant
2-Hydroxy-m-toluic acid
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Hydroxy-m-toluic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
83-40-9
:
Formula:
C₈H₈O₃
MDL Number:
MFCD00002448
Melting Pt:
163…167 °C
:
152.15 g/mol
EINECS:
201-473-8
:
GHS07
Storage Temperature:
Ambient