
Ethyl morpholine-2-carboxylate
Catalog # APOSOR40501-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:159.19 g/mol
Boiling Pt:103…105 °C/1mmHg
Storage Temperature:Ambient
UN:2735
ADR:8,III
:GHS05
CAS Number:135782-25-1
Formula:C₇H₁₃NO₃
MDL Number:MFCD08751336