Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Iodophenyl isocyanate
4-Iodophenyl isocyanate
4-Iodophenyl isocyanate
:
Apollo Scientific
4-Iodophenyl isocyanate
:
Apollo Scientific
:
15845-62-2
:
:
C₇H₄INO
:
MFCD00013875
:
245.02 g/mol
:
44…48 °C
:
113 °C
:
GHS07
:
Refrigerator