Order Entry
Finland
ContactUsLinkComponent
5-Bromo-3-pyridineboronic acid pinacol ester 97%
5-Bromo-3-pyridineboronic acid pinacol ester 97%
Catalog # APOSOR360813-5G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-3-pyridineboronic acid pinacol ester 97%
Catalog # APOSOR360813-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :283.96 g/mol
Storage Temperature:Ambient
CAS Number:452972-13-3
Formula:C₁₁H₁₅BBrNO₂
MDL Number:MFCD06798248