
4-(N,N-Dimethylamino)phenylboronic acid pinacol ester
Catalog # APOSOR360226-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:247.1410 g/mol
Melting Pt:124…126 °C
:GHS07
Storage Temperature:Ambient
CAS Number:171364-78-6
Formula:C₁₄H₂₂BNO₂
MDL Number:MFCD05663854