Order Entry
Finland
ContactUsLinkComponent
4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester
Irritant
4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester
Catalog # APOSOR346191-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Hydroxy-5-methoxy-2-nitrobenzoic acid methylester
Catalog # APOSOR346191-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.18 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:27883-60-9
Formula:C₉H₉NO₆
MDL Number:MFCD09833001