0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Finland
ContactUsLinkComponent
8-Acetyl-7-hydroxy-4-methylcoumarin
8-Acetyl-7-hydroxy-4-methylcoumarin
8-Acetyl-7-hydroxy-4-methylcoumarin
APOSOR345062-5G
:
Apollo Scientific
:
8-Acetyl-7-hydroxy-4-methylcoumarin
APOSOR345062-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
218.21 g/mol
:
170 °C
:
GHS07
:
Ambient
:
2555-29-5
:
C₁₂H₁₀O₄