Order Entry
Finland
ContactUsLinkComponent
3-Bromo-4-nitrobenzoic acid
Irritant
3-Bromo-4-nitrobenzoic acid
Catalog # APOSOR345005-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-4-nitrobenzoic acid
Catalog # APOSOR345005-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :246.02 g/mol
Melting Pt:200…204 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:101420-81-9
Formula:C₇H₄BrNO₄
MDL Number:MFCD04117949