Order Entry
Finland
ContactUsLinkComponent
 
Irritant
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%
Catalog # APOSOR321377-100G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
(2R,4R)-1-tert-Butyl-2-methyl-4-hydroxypyrrolidine-1,2-dicarboxylate 97%
Catalog # APOSOR321377-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :245.27 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:114676-69-6
Formula:C₁₁H₁₉NO₅
MDL Number:MFCD00797548