Order Entry
Finland
ContactUsLinkComponent
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate 97%
Irritant
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate 97%
Catalog # APOSOR321318-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate 97%
Catalog # APOSOR321318-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :169.18 g/mol
Melting Pt:54…56 °C
 :GHS07
Storage Temperature:Refrigerator
CAS Number:23286-70-6
Formula:C₇H₁₁N₃O₂
MDL Number:MFCD03453130