Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 98%
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 98%
:
Apollo Scientific
(1S,4R)-(+)-4-Acetoxy-2-cyclopentene-1-ol 98%
:
Apollo Scientific
:
60410-16-4
:
MFCD00210003
:
C₇H₁₀O₃
:
MFCD00078933
:
142.154 g/mol
:
43 °C
:
GHS07
:
Refrigerator