Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Bromo-2-nitroanisole
5-Bromo-2-nitroanisole
5-Bromo-2-nitroanisole
:
Apollo Scientific
5-Bromo-2-nitroanisole
:
Apollo Scientific
:
103966-66-1
:
:
C₇H₆BrNO₃
:
MFCD09966092
:
232.03 g/mol
:
90…92 °C
:
GHS07
:
Ambient