
1-Boc-piperidin-4-ylboronic acid pinacol ester 96%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1048970-17-7
:
Formula:C₁₆H₃₀BNO₄
MDL Number:MFCD09997792
:311.2247 g/mol
Melting Pt:60…65 °C
:GHS07
Storage Temperature:Refrigerator