Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Indazole-7-boronic acid pinacol ester
1H-Indazole-7-boronic acid pinacol ester 97%
1H-Indazole-7-boronic acid pinacol ester 97%
Irritant
1H-Indazole-7-boronic acid pinacol ester 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
1H-Indazole-7-boronic acid pinacol ester 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
915411-02-8
:
Formula:
C₁₃H₁₇BN₂O₂
MDL Number:
MFCD08059264
:
244.0973 g/mol
Melting Pt:
180…184 °C
:
GHS07
Storage Temperature:
Ambient