Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
Supplier:
Apollo Scientific
Certificates
Expand image
(1S,2S)-trans-2-Aminocyclopentanol hydrochloride
Supplier:
Apollo Scientific
About this item
CAS Number:
68327-04-8
:
Formula:
C₅H₁₂ClNO
MDL Number:
MFCD07370092
:
137.6079 g/mol
Storage Temperature:
Ambient