
2-Methyl-2H-indazole-5-carboxylic acid methyl ester 97+%
Catalog # APOSOR30723-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:190.2 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1092351-86-4
Formula:C₁₀H₁₀N₂O₂
MDL Number:MFCD11109406