Order Entry
Finland
ContactUsLinkComponent
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) 95+%
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) 95+%
Catalog # APOSOR307149-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Methylcyclohexanecarboxylic acid (cis and trans mixture) 95+%
Catalog # APOSOR307149-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :142.20 g/mol
Storage Temperature:Ambient
CAS Number:4331-54-8
Formula:C₈H₁₄O₂
MDL Number:MFCD00074909