
3-Bromo-2-nitroanisole 98%
Catalog # APOSOR30707-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.0314 g/mol
Boiling Pt:130…131 °C/3mmHg
Melting Pt:73…74 °C
:GHS07
Storage Temperature:Ambient
CAS Number:500298-30-6
Formula:C₇H₆BrNO₃
MDL Number:MFCD11101044