5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:365996-06-1
:
Formula:C₁₁H₁₅BrN₂O₂S
MDL Number:MFCD09951958
Melting Pt:94…97 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:319.23 g/mol