
3-(5-Chloropyridin-2-yl)-3-oxopropanenitrile
Catalog # APOSOR303687-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:180.59 g/mol
Melting Pt:121…123 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:1107053-87-1
Formula:C₈H₅ClN₂O
MDL Number:MFCD15526709