3-Aminopyrazine-2-carbaldehyde
Catalog # APOSOR302553-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:123.1100 g/mol
Storage Temperature:Ambient
CAS Number:32710-14-8
Formula:C5H5N3O
MDL Number:MFCD10696886