Order Entry
Finland
ContactUsLinkComponent
5-Nitro-3-pyrazolecarboxylic acid
Irritant
5-Nitro-3-pyrazolecarboxylic acid
Catalog # APOSOR301102-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
5-Nitro-3-pyrazolecarboxylic acid
Catalog # APOSOR301102-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :157.0800 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:198348-89-9
Formula:C4H3N3O4
MDL Number:MFCD00192356