6-Bromoindole 98%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 6-Bromo-1H-indole
- CAS Number:52415-29-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00238550
- Molecular Weight:196.05 g/mol
- Boiling Pt:70…75 °C/0.01mmHg
- Melting Pt:92…96 °C
- Storage Temperature:Ambient
3 Options Available Below
Product Details & Documents
Synonyms: 6-Bromo-1H-indole
- CAS Number:52415-29-9
- Formula:C₈H₆BrN
- MDL Number:MFCD00238550
- Molecular Weight:196.05 g/mol
- Boiling Pt:70…75 °C/0.01mmHg
- Melting Pt:92…96 °C
- Storage Temperature:Ambient



