Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Diphenyl sulphide
Diphenyl sulphide 98%
Diphenyl sulphide 98%
:
Apollo Scientific
Diphenyl sulphide 98%
:
Apollo Scientific
:
139-66-2
:
:
C₁₂H₁₀S
:
MFCD00003064
:
186.28 g/mol
:
296 °C
:
–40 °C
:
1.118 g/cm³ (25 °C)
:
>110 °C
:
GHS07
:
Ambient