3-Bromoimidazo[1,2-a]pyridine
Catalog # APOSOR23399-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Melting Pt:80…82 °C
:197.0300 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:4926-47-0
Formula:C₇H₅BrN₂
MDL Number:MFCD00014556