Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,5-Dimethylpyrrole-3-carboxylic acid
2,5-Dimethylpyrrole-3-carboxylic acid
2,5-Dimethylpyrrole-3-carboxylic acid
2,5-Dimethylpyrrole-3-carboxylic acid
Supplier:
Apollo Scientific
Certificates
Expand image
2,5-Dimethylpyrrole-3-carboxylic acid
Supplier:
Apollo Scientific
About this item
CAS Number:
57338-76-8
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD00203874
:
139.16 g/mol
Storage Temperature:
Ambient