Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Aminoquinoline
2-Aminoquinoline 96%
2-Aminoquinoline 96%
:
Apollo Scientific
2-Aminoquinoline 96%
:
Apollo Scientific
:
580-22-3
:
:
C₉H₈N₂
:
MFCD00038079
:
144.18
:
125…131 °C
:
209-458-8
:
GHS05
:
GHS07
:
Ambient