Order Entry
Finland
Orders LinkContactUsLinkComponent
 

3-Bromi-2-nitropyridiini 97+%

Irritant
Supplier:   Apollo Scientific
3-Bromi-2-nitropyridiini 97+%
Supplier:  Apollo Scientific
Certificates
3-Bromi-2-nitropyridiini 97+%

2 Options Available Below

Product Details & Documents
  • CAS Number:54231-33-3
  • Formula:C₅H₃BrN₂O₂
  • MDL Number:MFCD00955614
  • Molecular Weight:203 g/mol
  • Melting Pt:102…106 °C
  • Storage Temperature:Huoneenlämpö
Product Family Options

Product Information

  • Pakkaustyyppi
    Pak.
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 22,10 €
    22,10 €
    1 * 1 g
       22,10 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 32,40 €
    32,40 €
    1 * 5 g
       32,40 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.