Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Amino-4-methylpyridine
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine 98%
3-Amino-4-methylpyridine 98%
APOSOR2023-100G
:
Apollo Scientific
:
3-Amino-4-methylpyridine 98%
APOSOR2023-100G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
100 g
Seq:
3401
:
:
108.14 g/mol
:
254 °C
:
102…107 °C
:
GHS07
:
Ambient
:
3430-27-1
:
C₆H₈N₂
:
MFCD00128871