Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Butyne
1-Butyne
1-Butyne
:
Apollo Scientific
1-Butyne
:
Apollo Scientific
:
107-00-6
:
:
C₄H₆
:
54.09 g/mol
:
Refrigerator
:
2452
:
2.1,
:
GHS02
:
GHS04