
Cyclopropyl phenyl sulphide
Catalog # APOSOR18541-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:150.24 g/mol
Boiling Pt:62…63 °C/1mmHg
Density:1.045 g/cm³ (25 °C)
Flash Pt:92 °C
Storage Temperature:Ambient
UN:2810
ADR:6.1,III
:GHS06
CAS Number:14633-54-6
Formula:C₉H₁₀S
MDL Number:MFCD00009684