2-(4-Nitrophenyl)ethanol 98%
Catalog #
Product Details & Documents
Synonyms: 4-Nitrophenethyl alcohol, p-Nitrophenethyl Alcohol
- CAS Number:100-27-6
- Formula:C₈H₉NO₃
- MDL Number:MFCD00010202
- Molecular Weight:167.16 g/mol
- Boiling Pt:177 °C/16mmHg
- Melting Pt:59…62 °C
- Storage Temperature:Ambient
- EINECS:202-835-8



