
Methyl-3,4-dihydro-2H-1,4-benzoxazine-7-carboxylate
Catalog # APOSOR16468-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:193.2 g/mol
Melting Pt:86…88 °C
:GHS07
Storage Temperature:Ambient
CAS Number:142166-01-6
Formula:C₁₀H₁₁NO₃
MDL Number:MFCD11848180