
2-(2-Nitrophenyl)ethanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:15121-84-3
:
Formula:C₈H₉NO₃
MDL Number:MFCD00007192
:167.17 g/mol
Boiling Pt:267 °C
Melting Pt:2 °C
Density:1.19 g/cm³ (25 °C)
Flash Pt:113 °C
EINECS:239-178-1
:GHS07
Storage Temperature:Ambient