
2-Amino-3-bromo-5-nitrobenzonitrile 98%
Catalog # APOSOR1531-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:242.03 g/mol
Melting Pt:180…185 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:17601-94-4
Formula:C₇H₄BrN₃O₂
MDL Number:MFCD00054185