
1-Bromo-3-chlorobenzene 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108-37-2
:
Formula:C₆H₄BrCl
MDL Number:MFCD00000568
Boiling Pt:195…197 °C
Melting Pt:–22 °C
Density:1.63 g/cm³ (25 °C)
Flash Pt:80 °C
:191.45 g/mol
EINECS:203-575-8
:GHS07
Storage Temperature:Ambient