
N-(tert-Butoxycarbonyl)-3-(2-naphthyl)-D-alanine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76985-10-9
:
Formula:C₁₈H₂₁NO₄
MDL Number:MFCD00076900
:315.37 g/mol
Melting Pt:95…100 °C (dec.)
:GHS07
Storage Temperature:Ambient