Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Boc-2-chloro-L-phenylalanine
Boc-2-chloro-L-phenylalanine
Boc-2-chloro-L-phenylalanine
APOSOR14540-1G
:
Apollo Scientific
:
Boc-2-chloro-L-phenylalanine
APOSOR14540-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
299.76 g/mol
:
94…100 °C
:
GHS07
:
Ambient
:
114873-02-8
:
C₁₄H₁₈ClNO₄
:
MFCD00672514