
2-Chloro-4-methylpyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3678-62-4
:
Formula:C₆H₆ClN
MDL Number:MFCD00023418
:127.57 g/mol
Boiling Pt:194…195 °C
Density:1.142 g/cm³ (25 °C)
EINECS:222-951-2
:GHS07
Storage Temperature:Ambient