
3-(tert-Butoxycarbonyl)aminomethylphenylboronic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:199609-62-6
:
Formula:C₁₂H₁₈BNO₄
MDL Number:MFCD06246052
:251.09 g/mol
Melting Pt:188…190 °C
:GHS07
Storage Temperature:Refrigerator