
1-Boc-4-(cyanomethyl)piperidine
Catalog # APOSOR11986-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:224.3000 g/mol
Melting Pt:70…74 °C
:GHS07
Storage Temperature:Refrigerator
CAS Number:256411-39-9
Formula:C₁₂H₂₀N₂O₂
MDL Number:MFCD09801019