Order Entry
Finland
ContactUsLinkComponent
3-Bromo-5-nitrobenzonitrile 98%
Irritant
3-Bromo-5-nitrobenzonitrile 98%
Catalog # APOSOR11657-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-Bromo-5-nitrobenzonitrile 98%
Catalog # APOSOR11657-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :227.02 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:49674-15-9
Formula:C₇H₃BrN₂O₂
MDL Number:MFCD00463804