Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Bromo-4-nitrobenzoic acid
2-Bromo-4-nitrobenzoic acid 98%
2-Bromo-4-nitrobenzoic acid 98%
2-Bromo-4-nitrobenzoic acid 98%
Supplier:
Apollo Scientific
Certificates
Expand image
2-Bromo-4-nitrobenzoic acid 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
16426-64-5
:
Formula:
C₇H₄BrNO₄
MDL Number:
MFCD00234252
:
246.02 g/mol
Melting Pt:
165…168 °C
:
GHS07
Storage Temperature:
Ambient