
6-Chloroimidazo[1,2-a]pyridine hydrochloride 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:957035-24-4
:
Formula:C₇H₆Cl₂N₂
MDL Number:MFCD09475876
:189.0400 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together