
4,6-Dihydroxypyrimidine
Catalog # APOSOR11295-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:112.09 g/mol
Melting Pt:>300 °C
EINECS:214-772-3
:GHS07
Storage Temperature:Ambient
CAS Number:1193-24-4
Formula:C₄H₄N₂O₂
MDL Number:MFCD00016733
Melting Pt:>300 °C