Order Entry
Finland
ContactUsLinkComponent
4-Methyl-2-thenoic acid 97%
Irritant
4-Methyl-2-thenoic acid 97%
Catalog # APOSOR11275-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4-Methyl-2-thenoic acid 97%
Catalog # APOSOR11275-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
Melting Pt:122…126 °C
 :142.18 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:14282-78-1
Formula:C₆H₆O₂S
MDL Number:MFCD00089284