Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Bromobenzo[d]isoxazol-3-amine
6-Bromobenzo[d]isoxazol-3-amine 97%
6-Bromobenzo[d]isoxazol-3-amine 97%
APOSOR110302-1G
:
Apollo Scientific
:
6-Bromobenzo[d]isoxazol-3-amine 97%
APOSOR110302-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
213.0314 g/mol
:
114…116 °C
:
GHS07
:
Refrigerator
:
177995-39-0
:
C₇H₅BrN₂O
:
MFCD09788587